Applied Biosystems™

SOLiD™ FWD2 SP Kit

Catalog number:  4463012

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ FWD2 SP Kit contains sequencing primers and bridge probes for mate-paired sequencing runs, as well as multiplex fragment and multiplex paired-end sequencing runs. These are used to prime (1) the R3 tag of mate-paired libraries or (2) the barcode tag of multiplexed libraries for ligation sequencing in the forward direction.

The SOLiD™ Sequencing Primer (SP) Kits contain sequencing primers and other reagents needed for ligation sequencing on the 5500⁄5500xl Genetic Analyzer. The SOLiD™ FRD2 SP kit includes:
• one strip tube containing specific sequencing primers and bridge probes
• one SOLiD™ Sequencing QC strip tube
• four strip tubes of SOLiD™ Mixing Tubes

This kit contains sufficient primers to run 12 lanes.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Format: Strip Tubes
Product Line: SOLiD™
Product Size: 1 kit
Recommended Applications: De Novo Sequencing, Resequencing, Short PCR Product Sequencing, Shotgun Sequencing
Technique: Ligation-Based Sequencing
Template Compatibility: Barcodes (for Multiplexed Samples), Genomic DNA (Bacterial)
Shipping Condition: Dry Ice

Contents & storage

Store the kit at –25°C to –15°C until ready for use.

Documents

Manuals & protocols