SOLiD™ Buffer B, Double

Catalog number: 4463021

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ Buffer B, Double (PN 4463021) consists of SOLiD™ Buffer B Part 1 and SOLiD™ Buffer B Part 2, which need to be mixed prior to running on the instrument. SOLiD™ Buffer B, Double (PN 4463021) contains sufficient reagents to run 950 to 1632 cycles, or up to 136 cycles per lane.

The SOLiD™ Buffers are needed for ligation sequencing on the 5500⁄5500xl Genetic Analyzer. These kits are individually orderable components of the SOLiD™ Run Cycle Buffer Kit (PN 4461840). Supplemental buffers can be used to replace components of the main buffer kit. Each double size has sufficient buffer for covering two FlowChips.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: Applied Biosystems™, SOLiD™
Product Size: 1 kit
Recommended Applications: De Novo Sequencing, Shotgun Sequencing
Shipping Condition: Wet Ice

Contents & storage

Store SOLiD™ Buffer B components at 2°C to 8°C until ready to mix and use

Documents

Manuals & protocols