Applied Biosystems™

SOLiD™ REV Buffer, Double

Catalog number:  4463027

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ Buffers are used for ligation sequencing on the 5500⁄5500xl Genetic Analyzers. The SOLiD™ REV Buffer Kit (Double) is used during paired-end runs for ligation sequencing in the reverse direction. The SOLiD™ REV Buffer kit consists of two components (SOLiD™ REV Buffer Part 1 and SOLiD™ REV Buffer Part 2) that need to be mixed prior to use. The kit contains sufficient reagents to run 240 to 420 cycles, or up to 35 cycles in one lane.

The SOLiD™ REV Buffer is required only for paired-end sequencing runs.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: SOLiD™
Product Size: 1 kit
Recommended Applications: De Novo Sequencing, Shotgun Sequencing
Shipping Condition: Dry Ice

Contents & storage

This kit contains buffers for two 5500⁄5500xl FlowChips.

Store SOLiD™ REV Buffer Kit at 2°C to 8°C until ready for mixing and use.

Documents

Manuals & protocols