Calcium acetate monohydrate, ACS reagent
Calcium acetate monohydrate, ACS reagent
Calcium acetate monohydrate, ACS reagent
Thermo Scientific Chemicals

Calcium acetate monohydrate, ACS reagent

CAS: 5743-26-0 | C4H6CaO4·H2O | 176.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 446355000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5743-26-0
IUPAC Namecalcium diacetate hydrate
Molecular FormulaC4H8CaO5
InChI KeyXQKKWWCELHKGKB-UHFFFAOYSA-L
SMILESO.[Ca++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
AlkalinityPasses test
Heavy metals (as Pb)=<0.005 %
Iron (Fe)=<0.001 %
Strontium (Sr)=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only