Applied Biosystems™

AB Library Builder™ System, self installation

Catalog number:  4463592

 Related applications:

SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The AB Library Builder™ System simplifies next-generation sequencing by providing a semi-automated solution for DNA fragment library preparation for the SOLiD™ 4 System and 5500 Series Genetic Analysis Systems. The AB Library Builder™ System is designed to increase throughput and reduce labor associated with library preparation for exome and targeted resequencing applications. The workflow can be simplified even further by using the system for upstream DNA purification. Automated—reducing hands-on time by 50%.
Purchased for self installation (PN4463592).

• Scalable: Prepares up to 13 libraries per run, up to 26 libraries per day
• Integrated: A complete system optimized for library preparation for the SOLiD™ 4 System and 5500 Series Genetic Analysis Systems.
• Automated Workflow: The AB Library Builder™ System automates the most labor-intensive parts of DNA fragment library preparation.

The AB Library Builder™ System addresses low- to medium-throughput needs for DNA fragment libraries for exome and targeted resequencing applications. It can process up to 13 samples at a time, and with a second run can yield up to 26 libraries in a day. In the time needed to prepare 8 libraries manually, the AB Library Builder™ System can prepare 13 libraries.

Reagents kits for DNA fragment library creation using the AB Library Builder™ are available for purchase separately. These kits require minimal set-up and come with ready to use reagent cartridges for the creation of up to 13 DNA fragment libraries. Oligos, Barcode adaptors, and Agilent AMPure XP beads are also purchased separately.
The AB Library Builder™ System provides full walk-away automation of the post-shearing and pre-nick translation portion of the DNA fragment library workflow for exome and targeted resequencing.

Note: For Research Use Only. Not intended for animal or human therapeutic or diagnostic use
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Up to 13 libraries per run
For Use With (Equipment): 5500 Genetic Analyzer, Ion PGM™ System, SOLiD™ 4 System
Library Type: Fragment Library, cDNA Library
Product Line: Library Builder™
Product Size: 1 instrument

Documents

Manuals & protocols