AB Library Builder™ Tip and Tube Rack

Catalog number:  4463776

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The AB Library Builder™ Tip and Tube Rack is an accessory that is used with the AB Library Builder™ System. It can be ordered separately as a replacement to the Tip and Tube Rack that comes with a system purchase. Also, additional racks can be purchased to allow processing of multiple batch runs.

The AB Library Builder™ Tip and Tube Rack can hold enough sample tubes, tips, and tip holders for the creation of up to 13 libraries per run using the AB library Builder™ System.
For Research Use Only. Not for use in diagnostic procedures.



For Use With (Equipment): AB Library Builder™
Product Line: Library Builder™
Product Size: 1 unit
Shipping Condition: Room Temperature
Type: Rack

Contents & storage

Store at room temperature.


Manuals & protocols