Calcium sulfate dihydrate, 98+%, extra pure
Calcium sulfate dihydrate, 98+%, extra pure
Calcium sulfate dihydrate, 98+%, extra pure
Calcium sulfate dihydrate, 98+%, extra pure
Thermo Scientific Chemicals

Calcium sulfate dihydrate, 98+%, extra pure

CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 446380050
Price (EUR)
218,00
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
218,00
Each
Add to cart
Chemical Identifiers
CAS10101-41-4
IUPAC Namecalcium dihydrate sulfate
Molecular FormulaCaH4O6S
InChI KeyPASHVRUKOFIRIK-UHFFFAOYSA-L
SMILESO.O.[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric>=98.0 %
Loss on ignition18 to 22 % (800°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only