5500 Series Genetic Analyzers UPS (Uninterruptable Power Supply)

Catalog number: 4463927

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is an Uninterruptible Power Supply (UPS) unit to safeguard the SOLiD 4 and 5500 series Genetic Analysis Systems.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Product Line: SOLiD™
Product Size: 1 unit
Shipping Condition: Room Temperature
Type: Power Supply

Contents & storage

Store at room temperature.

Documents

Manuals & protocols