Applied Biosystems™

5500 SOLiD™ Fragment Library Core Kit

Catalog number:  4464412

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 SOLiD™ Fragment Library Core Kit is used to construct fragment libraries for sequencing on the 5500 Series Genetic Analysis Systems.

The 5500 SOLiD™ Fragment Library Core Kit contains the necessary reagents (enzymes, buffers, and PCR Amplification mix) for preparing 12 fragment libraries. Use in conjunction with either the 5500 SOLiD™ Fragment Library Standard Adaptor kit (PN 4464411) or the appropriate 5500 SOLiD™ Fragment Library Barcode Adaptor kit, which should be ordered separately.

The Core Kit consists of the 5500 SOLiD™ Fragment Library Enzyme Module (PN 4464413) and the 5500 SOLiD™ Fragment Library Amplification Module (PN 4464414).

Fragment libraries consist of short lengths of DNA specifically selected for optimal performance with a next generation sequencing platform for applications such as targeted or whole genome resequencing. Briefly, the 5500 SOLiD™ fragment library preparation protocol takes DNA through shearing, end-repair, A-tailing, and ligation with only one intervening bead-based purification that doubles as a size selection step. In many cases, subsequent library amplification is not needed, allowing high complexity libraries to be brought directly into the clonal amplification step.

Fragment libraries can be prepared in about four hours (or about five hours with amplification).

Key benefits:
• Optimized set of reagents for construction of 12 fragment libraries for use with the 5500 Series Genetic Analysis Systems
• Enable the use of sequencing control (with presence of an internal adaptor)
• Support amplification-free workflow (depending on desired yield)
• Bead-based cleanup for faster and more efficient processing
• Lower cost per library (compared to SOLiD™ fragment library kits)

The 5500 SOLiD™ Fragment Library Core Kit is specifically optimized for use with the 5500 Series Genetic Analysis Systems.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Library Type: Fragment Library
Workflow Step: Library Barcoding, Library Generation
Starting Material: DNA
Sequencing Type: Genome & DNA Sequencing
Downstream Application: Next-Generation Sequencing
Final Product: Fragment Library
Product Line: SOLiD™
Product Size: 1 kit
Recommended Applications: Small Genome Sequencing, Targeted Resequencing

Contents & storage

The kit contains a Fragment Library Enzyme Module and a Fragment Library Amplification Module. Store both modules at -20°C. Each kit contains sufficient material for constructing 12 fragment libraries.

Documents

Manuals & protocols