Applied Biosystems™

AB Library Builder™ Builder D-Ring Tool

Catalog number:  4465603

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The AB Library Builder™ D-Ring Tool is an accessory that is used for removal and replacement of the AB Library Builder™ D-Rings. It can be ordered separately as a replacement to the D-Ring Tool that comes with a system purchase
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): AB Library Builder™
Product Line: Library Builder™
Product Size: 1 unit
Shipping Condition: Room Temperature
Type: Other Accessories

Contents & storage

One D-Ring tool

Documents

Manuals & protocols