Attune® Acoustic Focusing Cytometer Emission Filter Holder Blade

Catalog number: 4465832

Applied Biosystems™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This emission filter holder can be used in combination with an optical filter to develop custom filter sets for the Attune® Acoustic Focusing Cytometer. Filter not included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Line: Attune®
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Optical Accessories

Contents & storage

Store at room temperature

Documents

Manuals & protocols