Applied Biosystems™

Attune™ Acoustic Focusing Cytometer Dichroic Filter Holder Blade

Catalog number:  4465834

 Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This dichroic filter holder can be used in combination with a dichroic filter to develop custom filter sets for the Attune® Acoustic Focusing Cytometer. Filter not included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Line: Attune™
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Optical Accessories

Contents & storage

Store at room temperature

Documents

Manuals & protocols