Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™
Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™
Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™
Thermo Scientific Chemicals

Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™

CAS: 1907-33-1 | C4H9LiO | 80.05 g/mol
数量:
100 mL
800 mL
製品番号(カタログ番号) 446848000
価格(JPY)
-
数量:
800 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1907-33-1
IUPAC Namelithium(1+) 2-methylpropan-2-olate
Molecular FormulaC4H9LiO
InChI KeyLZWQNOHZMQIFBX-UHFFFAOYSA-N
SMILES[Li+].CC(C)(C)[O-]
さらに表示
Strength0.80 to 1.10 M
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Clear yellow to brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only