Acetic acid, sodium salt, 99%, for HPLC, anhydrous
Acetic acid, sodium salt, 99%, for HPLC, anhydrous
Acetic acid, sodium salt, 99%, for HPLC, anhydrous
Thermo Scientific Chemicals

Acetic acid, sodium salt, 99%, for HPLC, anhydrous

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 446880500
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.20
Save 8.55
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.20
Save 8.55
Each
Chemical Identifiers
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 %
UVat 260 nm A: =<0.02
UV(0.5 M in water)
Loss on drying=<1 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only