POROS™ CaptureSelect™ LC Lambda Affinty HPLC Columns
Thermo Scientific™

POROS™ CaptureSelect™ LC Lambda Affinty HPLC Columns

Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoLongitud (métrico)Diámetro (métrico)
446916350 mm4.6 mm
446915030 mm2.1 mm
4469168100 mm4.6 mm
Número de catálogo 4469163
Precio (MXN)
-
Longitud (métrico):
50 mm
Diámetro (métrico):
4.6 mm

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

POROS CaptureSelect LC Lambda HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three minute quantitation of Fabs, immunoglobulins, and bispecific antibodies having a Lambda light chain.

Features:

  • Broad utility—POROS A, G and CaptureSelectaffinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins and fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—deliver high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect LC Lambda affinity ligands when combined with POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid quantitation and small-scale purification of all immunoglobulins and Fabs having a Lambda light chain (all subclasses).

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS Affinity Chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

LC Lambda Column Specifications:

  • Protein Binding Capacity: 10 mg/mL
  • Species Specificity: Human
  • Column Material: PEEK™ polymer or stainless steel
  • Frit: 2 μm, titanium
  • POROS Resin: Cross-linked poly(styrenedivinylbenzene)
  • CaptureSelect Ligands: Single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae
For Research Use Only. Not for use in diagnostics procedures.
Especificaciones
Capacidad de unión10.0 mg/mL IgG
Tipo de columnaAffinity
Volumen de elución máx1.6 mL
Volumen de elución mín800 μL
CaudalUp to 5000 cm/hr.
Para utilizar con (aplicación)HPLC
Compatibilidad de alto rendimientoNot High-throughput Compatible (Manual)
IncluyeColumn includes EZ grip fittings and column test certificate.
MatrizPolystyrenedivinylbenzene
Presión máxima180 bar
Tipo de productoColumn
Cantidad1 columna
ReactividadHuman
Tipo de muestraCell Culture Supernatants
Condiciones de envíoWet Ice
Fase estacionariaLight Chain Lambda affinity ligand, human-specific
Molécula dianaLambda Light Chain
Volumen (métrico)0.8 mL
Diámetro (métrico)4.6 mm
Longitud (métrico)50 mm
MaterialPEEK
Tamaño de partículas20 μm
FaseAffinity Separation
Tamaño de poro2000 Å
Línea de productosCaptureSelect, POROS
Diana de purificaciónLambda Light Chain
TécnicaAntibody Affinity Chromatography
Unit SizeEach

Preguntas frecuentes

Are POROS analytical columns compatible with HPLC systems from Waters or Agilent?

All of the pre-packed POROS columns, regardless of column size or resin, have the same 10-32 female fittings on the column. Columns are compatible with Waters and Agilent HPLC systems. You will need 1/16 inch male fittings for the HPLC to attach the column.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

For any POROS analytical column, is the column compatible with an AKTA or Avant purification system?

All of the pre-packed columns, regardless column size or resin, have the same 10-32 female fittings on the column. They are all compatible with the AKTA instrument platform, however due to the column volume of the 2.1 mmD x 30 mmL columns (0.1 mL), these will not perform well in AKTA or Avant purification systems due to the relative size of the hold- up volume in these systems. The smallest column (PEEK or SS) that could feasibly be used in these systems is the 4.6mmD x 50mmL (0.8 mL) format.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Are the pre-packed POROS CaptureSelect or any POROS analytical columns compatible with an UPLC or is the use of these columns limited to HPLC?

UPLC is typically described as chromatography with sub-2 µm particles, which drives higher efficiency/better separation independent of operating flow rate. As particle size decreases, backpressure increases so the UPLC systems tend to have higher pressure ratings compared to HPLC systems. The flow rates on UPLC systems would be sufficient for POROS columns. Pump capabilities are typically 0.1-1.0 mL/min, so not as fast as a HPLC, but sufficient. Hold-up volumes on UPLC systems are lower than HPLCs, so this can help with sensitivity and efficiency. So from a pressure and flow standpoint there should be no challenge to operate a POROS CaptureSelect column on a UPLC system.

That being said, care should be taken running POROS columns on UPLC systems. Higher pressure safety interlock MUST be set at the rated column pressure, typically 180 bar. HPLC column hardware is not rated to operate at the static pressures possible and typical on UPLC systems. A clog or the like on a UPLC system could generate unsafe pressures on any HPLC-rated column before the default UPLC safety pressure interlock is tripped.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the physical or mechanical stability of POROS resins?

The physical stability of POROS resins is high. Customers have agitated, with overhead mixing and or bottom gassing for long periods of time (5-10 hours) and have performed hundreds of diaphragm pump passes during stall packs. Keep in mind that agitation with any device that creates grinding and sheer, such as with a magnetic stir bar, is not recommended for any type of resin.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What should I do if a POROS column dries out or air is pumped onto the column?

POROS resins rehydrate quite well. Here is the procedure that we recommend:

1. Flow 5% ethanol over the column at ?350 cm/hour for 3 CVs, followed by water or 0.1 M sodium chloride at 300 cm/hour for 3 CVs.
2. If baseline noise is still present, flow 20% ethanol over the column at 300 cm/hour for 3 CVs, followed by water or 0.1 M sodium chloride at 300 cm/hour for 3 CVs.
3. If you have established column qualification tests and specifications, perform the tests after rewetting.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.