Ammonium dichromate, ACS reagent
Ammonium dichromate, ACS reagent
Ammonium dichromate, ACS reagent
Thermo Scientific Chemicals

Ammonium dichromate, ACS reagent

CAS: 7789-09-5 | H8Cr2N2O7 | 252.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 446921000
Price (EUR)
34,30
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
34,30
Each
Chemical Identifiers
CAS7789-09-5
IUPAC Namediammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2H8N2O7
InChI KeyJOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Appearance (Form)Crystalline powder
Loss on drying=<3.0 %
Chloride (Cl)=<0.005 %
Calcium (Ca)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only