Sodium oxalate, ACS reagent
Sodium oxalate, ACS reagent
Sodium oxalate, ACS reagent
Thermo Scientific Chemicals

Sodium oxalate, ACS reagent

CAS: 62-76-0 ∣ C2Na2O4 ∣ 134.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 446961000
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.50
Save 9.85
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.50
Save 9.85
Each
Chemical Identifiers
CAS62-76-0
IUPAC Namedisodium oxalate
Molecular FormulaC2Na2O4
InChI KeyZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.01 %
Ammonium (NH4)=<0.002 %
Heavy metals (as Pb)=<0.002 %
Appearance (Form)Crystalline powder
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only