Product Image
Applied Biosystems™

QuantStudio™ 12K Flex OpenArray™ Practice Kit

The QuantStudio 12K Flex OpenArray Practice Kit is designed to introduce newcomers to the OpenArray block workflow, as well asRead more
Have Questions?
Catalog NumberQuantity
44696201 kit
Catalog number 4469620
Price (USD)
2,655.00
Each
Quantity:
1 kit
Price (USD)
2,655.00
Each
The QuantStudio 12K Flex OpenArray Practice Kit is designed to introduce newcomers to the OpenArray block workflow, as well as to provide instrument verification. The kit contains 6 practice loading arrays plus reagents. The practice kit also includes these accessories: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill tips, and 384-well plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™ 12k Flex
Format384-Well Plate
Genotyping TargetSNPs (Known)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineOpenArray, QuantStudio
Product Type12K Flex Practice Kit
Quantity1 kit
Sample TypeDNA
TechniqueSNP Genotyping (Real-Time PCR-Based)
Detection MethodPrimer-probe
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
The practice kit includes a QuantStudio Accessories Kit, Accufill System tips, 384-well sample plates and loading plate.

Frequently asked questions (FAQs)

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.