What applications can I perform using digital PCR?
Digital PCR can be used to perform rare allele detection, absolute quantitation of gene expression, viral titering, and absolute quantitation of next-generation sequencing libraries.
Search
View additional product information for QuantStudio™ Digital PCR Kit - FAQs (4470185)
5 product FAQs found