1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™
1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™
1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™
Thermo Scientific Chemicals

1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 447060025
Price (USD)
-
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.01 %
Water=<0.1 %
Heavy metals=<5 ppm
Peroxides=<0.005 % (as H2O2)
UVat 250 nm A: =<0.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only