Product Image
Applied Biosystems™
QuantStudio™ 12K Flex OpenArray™ Block Upgrade Kit (with Accufill™ System)
The QuantStudio™ 12K Flex OpenArray™ Block Upgrade Kit (with Accufill™ System) can accelerate genomic validation and screening, driving over 150,000Más información
Have Questions?
Número de catálogoCantidad
44710671 block
Número de catálogo 4471067
Precio (CLP)
-
Cantidad:
1 block
The QuantStudio™ 12K Flex OpenArray™ Block Upgrade Kit (with Accufill™ System) can accelerate genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.
Plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the Accufill™. Easily load 4 arrays into the QuantStudio 12K Flex instrument to achieve over 12,000 data points.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Especificaciones
Para utilizar con (aplicación)PCR en tiempo real (PCRq)
Para utilizar con (equipo)QuantStudio™
FormatoOpenArray™ Plate
Compatibilidad de placasOpenArray™ Plate
Línea de productosQuantStudio
Cantidad1 block
Tipo de productoBlock Upgrade Kit
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente
Preguntas frecuentes
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?
Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.