Product Image
Applied Biosystems™

TaqMan™ OpenArray™ Loading Plate, QuantStudio™ 12K Flex

The TaqMan™ OpenArray™ Loading Plate, QuantStudio™ 12K Flex is designed to provide new and experienced users a single OpenArray™ plateRead more
Have Questions?
Catalog NumberQuantity
44712271 Plate
Catalog number 4471227
Price (USD)
748.00
Each
Quantity:
1 Plate
Price (USD)
748.00
Each
The TaqMan™ OpenArray™ Loading Plate, QuantStudio™ 12K Flex is designed to provide new and experienced users a single OpenArray™ plate to practice the workflow prior to starting custom projects.

The OpenArray™ loading plate does not contain assays and is used for practicing loading techniques.

For Research Use Only. Not for use in diagnostics procedures.
Specifications
For Use With (Equipment)QuantStudio™ 12k Flex
FormatOpenArray Plate
Product LineOpenArray, TaqMan
Product TypeLoading Plate
Quantity1 Plate
Detection MethodPrimer-probe
For Use With (Application)Real Time PCR (qPCR)
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
1 plate., store at -20°C

Frequently asked questions (FAQs)

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.