Ammonium sulfate, 99%, for biochemistry
Ammonium sulfate, 99%, for biochemistry
Ammonium sulfate, 99%, for biochemistry
Thermo Scientific Chemicals

Ammonium sulfate, 99%, for biochemistry

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
製品番号(カタログ番号) 447135000
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
さらに表示
Titration excess NaOH>=98.5 %
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<5 ppm
Arsenic (As)=<1 ppm
Cadmium (Cd)=<1 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only