Potassium chromate, ACS reagent, meets the requirements of Reag.Ph.Eur.
Potassium chromate, ACS reagent, meets the requirements of Reag.Ph.Eur.
Potassium chromate, ACS reagent, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals

Potassium chromate, ACS reagent, meets the requirements of Reag.Ph.Eur.

CAS: 7789-00-6 | CrK2O4 | 194.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
447201000100 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447201000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-00-6
IUPAC Namedipotassium dioxochromiumbis(olate)
Molecular FormulaCrK2O4
InChI KeyXMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH8.6 to 9.8 (5% soln. at 25°C)
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.03 %
Calcium (Ca)=<0.005 %
Sodium (Na)=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only