Thermo Scientific Chemicals

Potassium permanganate, ACS reagent, low in mercury

Catalog number: 447215000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium permanganate, ACS reagent, low in mercury

Catalog number: 447215000
500 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
7722-64-7
IUPAC Name
potassium manganesoylolate
Molecular Formula
KMnO4
InChI Key
VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES
[K+].[O-][Mn](=O)(=O)=O
Appearance (Color)
Dark green to brown-black
Appearance (Form)
Crystalline powder
Insoluble matter
=<0.2 %
Chloride and Chlorate (as Cl)
=<0.005 %
Sulfate (SO4)
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text