Protocol Card: Library Builder™ WT: RNA

Catalog number: 4472736

Applied Biosystems™  Related applications: RNA Sequencing | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is the protocol card for running the Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems on the AB Library Builder™ System. This protocol automates the creation of up to 13 RNA whole transcriptome libraries on the AB Library Builder™ System.

• Protocol card for AB Library Builder™ System
• RNA whole transcriptome libraries can be run on the SOLiD® 4 System and 5500 Series Genetic Analysis Systems

Labor Saving Automation
The Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems when used with the AB Library Builder™ System automates the most labor-intensive parts of RNA whole transcriptome library preparation, helping to reduce hands-on time by at least 50%. The Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems contains enough reagents and materials to process up to 13 samples at a time.

Integrated RNA Library Preparation
The Library Builder™ Whole Transcriptome Core Kit for 5500 Genetic Analysis Systems, along with the AB Library Builder™ System, is an integrated solution comprising a predefined software protocol, plug-and-play cartridge format reagents, and instrument. These kits are all optimized and validated for RNA purification and RNA whole transcriptome library preparation with the 5500 and SOLiD® 4.

To further maximize automation consider using RNA purification protocols with the iPrep™ PureLink® Total RNA Kit, compatible with the Library Builder™ System.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

For Use With (Equipment): AB Library Builder™
Format: Card
Product Line: Library Builder™
Product Size: 1 card
Shipping Condition: Room Temperature
Type: Card

Contents & storage

One card, store at room temperature

Documents

Manuals & protocols

Recommended products