Applied Biosystems™

SOLiD™ Pre Deposition Plus Kit Box 2

Catalog number:  4472962

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SOLiD® Pre Deposition Plus Kit Box 2 buffer components of the SOLiD® Pre Deposition Plus Kit. Box 2 contains TEX buffer and Low TE buffer.

The full SOLiD® Pre Deposition Plus Kit is also available for purchase, and contains both Box 1 and Box 2.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer, SOLiD™ 3 Plus System, SOLiD™ 4 System
Product Line: SOLiD™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

The SOLiD® Pre Deposition Plus Kit Box 2 is comprised of a single box.

Box 2 — Store at ambient temperature:
• TEX buffer
• Low TE buffer

Documents

Manuals & protocols