SOLiD® Pre Deposition Plus Kit

Catalog number: 4472967

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SOLiD® Pre Deposition Plus Kit features all the necessary components for depositing templated beads onto a slide for ligation sequencing, along with reagents for performing a 3'-end modification reaction on templated beads prior to deposition. This kit is fully compatible with the 5500 and 5500xl Genetic Analyzers, as well as the SOLiD® 4 System.

The SOLiD® Pre Deposition Plus Kit contains sufficient material for these EZ Bead™ System scale outputs:

Multiple E10 (125M beads)/E20 (250M beads) scale runs

• Or five E80 (1B beads) scale runs
• Or two E120 (2.2B beads) scale runs

SOLiD® Pre Deposition Plus Kits are scalable and flexible. Each kit works with templated beads prepared at any scale using the SOLiD™ EZ Bead™ System or manual methods. Each kit supports fragment, whole transcriptome, and mate-paired libraries.

Each SOLiD® Pre Deposition Plus Kit is comprised of two boxes. The first box contains terminal transferase enzyme, terminal transferase buffer, cobalt chloride solution, and bead linker. The second box holds TEX buffer and Low TE buffer. Box 1 requires storage at -15°C to -25°C. Box 2 can be stored at ambient temperature.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer, SOLiD™ 3 Plus System, SOLiD™ 4 System
Product Line: SOLiD™
Product Size: 1 kit

Contents & storage

Box 1 (store at -15°C to -25°C):
• SOLiD® 10X Cobalt Chloride
• SOLiD® Terminal Transferase Buffer
• SOLiD® Bead Linker
• SOLiD® Terminal Transferase Enzyme

Box 1 (store at room temperature):
• TEX Buffer
• Low TE Buffer

Documents

Manuals & protocols