Product Image
Applied Biosystems™
SOLiD™ EZ Bead™ E120 System Consumables, for TargetSeq™-enriched samples
Have Questions?
Número de catálogoCantidad
44730004 packs
Número de catálogo 4473000
Precio (MXN)
-
Cantidad:
4 packs
SOLiD™ EZ Bead™ E120 System Consumables for TargetSeq™, 4-pack, contains all necessary reagents and consumables for performing four E120 EZ Bead™ system runs for TargetSeq™-enriched samples, except the SOLiD™ EZ Bead™ Oil Pack Kit, which can be purchased separately.
Included in the kit:
4 SOLiD™ EZ Bead™ Emulsifier E120 Reagents Kits
4 SOLiD™ EZ Bead™ Emulsifier E120 Accessories Kits
2 SOLiD™ EZ Bead™ Amplifier E120 Accessories Kits
4 SOLiD™ EZ Bead™ Enricher Buffer Reagents Kits
4 SOLiD™ EZ Bead™ Enricher E120 Reagents Kits
4 SOLiD™ EZ Bead™ Enricher Accessories Kits
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (aplicación)Sequencing, Resequencing
Para utilizar con (equipo)5500xl Genetic Analyzer, 5500 Genetic Analyzer, SOLiD™ 4 System
Línea de productosEZ Bead
Tipo de productoE120 System Consumables
Cantidad4 packs
Paso del flujo de trabajoBead Enrichment, Emulsion PCR
Unit Size4 packs
Contenido y almacenamiento
room temperature, 2 to 8°C and -15 to -25°C