SOLiD™ FWD SR S75 Kit, Single

Catalog number: 4473598

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD® Sequencing Reagents (SR) Kits contain enzymes and probes needed for ligation sequencing with 2-base encoded (2BE) probes on the 5500/5500xl Genetic Analyzer. The SOLiD® FWD SR Kits are used for fragment, mate-paired, and paired-end sequencing runs, for ligation sequencing in the forward direction.

Single-sized kits have sufficient reagents for covering one FlowChip.

The SOLiD® FWD SR S75 Kit, Single contains sufficient ligase, phosphatase, and forward 2BE probes to run 375 to 516 cycles, or up to 86 cycles per lane for 6 lanes. The kit is best configured for sequencing read lengths up to 75 bp for barcoded runs. In addition to the SOLiD® Run Cycle Buffers and SOLiD® Buffer D (Reset), this kit requires the use of the SOLiD® FWD Buffer. Store the SOLiD® FWD SR S75 Kits at –25°C to –15°C until ready for use.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Recommended Applications: De Novo Sequencing - High Throughput (cDNA), Resequencing, Short Read Sequencing, Standard Sequencing
Product Line: SOLiD™
Product Size: 1 kit
Shipping Condition: Dry Ice

Contents & storage

This kit contains probes and enzymes for one 5500⁄5500xl FlowChip.



Store the SOLiD™ FWD SR S75 Kit, Single at –25 °C to –15 °C until ready for use.

Documents

Manuals & protocols