Applied Biosystems™

SOLiD™ FWD SR S50 Kit, Double

Catalog number:  4473606

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD® Sequencing Reagents (SR) Kits contain enzymes and probes needed for ligation sequencing with 2-base encoded (2BE) probes on the 5500/5500xl Genetic Analyzer. The SOLiD® FWD SR Kits are used for fragment, mate-paired, and paired-end sequencing runs, for ligation sequencing in the forward direction.

Double-sized kits have sufficient reagents for covering two FlowChips.

The SOLiD® FWD SR S50 Kit, Double contains sufficient ligase, phosphatase, and forward 2BE probes to run 500 to 732 cycles, or up to 61 cycles per lane for 12 lanes. The kit is best configured for sequencing read lengths up to 50 bp for barcoded runs and sequencing read lengths up to 60 bp with no barcodes. In addition to the SOLiD® Run Cycle Buffers and SOLiD® Buffer D (Reset), this kit requires the use of the SOLiD® FWD Buffer. Store the SOLiD® FWD SR S50 Kits at –25°C to –15°C until ready for use.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Recommended Applications: De Novo Sequencing - High Throughput (cDNA), Resequencing, Short Read Sequencing, Standard Sequencing
Product Line: SOLiD™
Product Size: 1 kit

Contents & storage

This kit contains probes and enzymes for two 5500⁄5500xl FlowChips.



Store the SOLiD™ FWD SR S50 Kit, Double at –25 °C to –15 °C until ready for use.

Documents

Manuals & protocols