This product will be discontinued on March 1, 2017, and replaced by the NUCLEIC-CARD™ Collection Device System (available now). View notice »

NUCLEIC-CARD™ Sample Collection Device

Catalog number:  4473980

 Related applications: DNA Extraction | Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The NUCLEIC-CARD™ Sample Collection Device consists of a plastic holder carrying a framed NUCLEIC-CARD™ COLOR (1 spot). The NUCLEIC-CARD™ COLOR matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD™ COLOR changes color upon sample application to facilitate processing of buccal samples.

The design of the NUCLEIC-CARD™ Sample Collection Device enables accurate positioning of a 4N6FLOQSwabs™ Lollipop swab to ensure more efficient and uniform transfer of buccal cells with a few easy steps.

Key Features:

• Uniform and more reproducible sample transfer
• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems® direct PCR amplification kits
• Certified free of DNase, RNase, and amplifiable human DNA
• Prevents growth of microorganisms to enable long-term room temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity

The NUCLEIC-CARD™ Sample Collection Device is manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.

NUCLEIC-CARD™ is a trademark of Copan Flock Technologies.
For Forensic or Paternity Use Only.

Specifications

For Use With (Equipment): BSD Robotics Punching System
Format: Card
Product Line: NUCLEIC-CARD
Product Size: Case of 100
Shipping Condition: Room Temperature
Type: Sample Proccesser

Contents & storage

100 NUCLEIC-CARD Sample Collection Devices
Store at RT

Documents

Manuals & protocols

Recommended products