4N6FLOQSwabs™ Lollipop

Catalog number:  4473997

 Related applications: Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

4N6FLOQSwabs Lollipop have a solid molded plastic applicator shaft with a round-shaped tip to improve collection of buccal cells from the inside of a subject’s check. The tip of the 4N6FLOQSwabs Lollipop are coated with short Nylon fibers arranged in a perpendicular fashion to create a highly absorbent thin layer with an open structure. The perpendicular Nylon fibers act like a soft brush that facilitates improved collection of cellular material. Capillary action between the fiber strands facilitates strong hydraulic uptake of liquid sample, and the sample stays close to the surface for easy elution.

4N6FLOQSwabs Lollipop are specifically designed to improve buccal cell collection and increase transfer efficiency with the NUCLEIC-CARD Sample Collection Device.

4N6FLOQSwabs Lollipop are certified free of DNase, Rnase, and amplifiable human DNA.

4N6FLOQSwabs Lollipop are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
For Forensic or Paternity Use Only.

Specifications

Product Line: 4N6FLOQSwabs
Product Size: Case of 50
Shipping Condition: Room Temperature

Contents & storage

50 Lollipop FLOQSwabs, in peelpouch
Store at RT

Documents

Manuals & protocols