Elga® PURELAB® Flex Micron Filter, point of use

Catalog number: 4474532

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Point of Use (POU) Micron Filter for Small Option and MEDICA.

Excellent for ensuring consistent ultra pure water quality with respect to particles and bacteria immediately prior to dispensing.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

For Use With (Equipment): Ion PGM™ System
Product Line: Elga® PURELAB®
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Water Purification Accessories

Contents & storage

Store at room temperature

Documents

Manuals & protocols