Elga® PURELAB® Flex 3 Reverse Osmosis Module

Catalog number: 4474536

Ion Torrent™  Related applications: Ion Torrent™ Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Reverse Osmosis Cartridge for the Elga® PURELAB® flex 3.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

For Use With (Equipment): Ion PGM™ System
Product Line: Elga® PURELAB®
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Water Purification Accessories

Contents & storage

Store at room temperature

Documents

Manuals & protocols