Product Image
Applied Biosystems™
GeneMapper™ Software 6, client installation
Note: This client version of GeneMapper Software 6 installs the client configuration (user interface) only. Users must install GeneMapper SoftwareRead more
Have Questions?
Catalog NumberQuantity
44750741 License
Catalog number 4475074
Price (USD)
-
Quantity:
1 License
Note: This client version of GeneMapper Software 6 installs the client configuration (user interface) only. Users must install GeneMapper Software 6, full installation, prior to installing the client version, which should be installed on a computer separate from that of the full version.
GeneMapper Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all of our electrophoresis-based genotyping systems. This software specializes in multi-application functionality, including amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellite and SNP genotyping analysis. GeneMapper Software can help users increase data processing efficiency with remote auto-analysis and command line operation, and allows for multiuser, client-server deployment. The software uses Process Quality Values (PQVs) for automated identification that reduces data review time for high throughput genotyping. In addition, the security and audit features help users meet 21 CFR 11 requirements.
Features include:
• Installation on the data collection computer or a non-instrument computer
• Support of networked multiple clients when installed on a non-instrument computer
• An Oracle™ 11g database with project-related data storage that ensures faster installation times and latest database support, as well as flexible archive capabilities
• Customizable project auto-saving frequency
• Defining of linearity range in the analysis methods for more reproducible, higher quality sizing of larger fragments (>800bps)
• Analysis of 6-dye data generated from the Applied Biosystems 3500 and 3500xL instrument systems for higher multiplexing capabilities
GeneMapper Software has not undergone specific developmental validation for human identification applications. Human identification laboratories analyzing single-source or parentage samples that choose to use GeneMapper Software for data analysis should perform their own developmental validation studies.
Several users may access the database concurrently; however, because genotype analysis is data-intensive, more than five clients simultaneously accessing the database may slow the performance of GeneMapper Software.
Recommended Computer Configuration: Dell OptiPlex XE2, 7-4770S Intel Processor (Quad Core HT, 3.10GHz Turbo, 8MB), 16GB (2x8GB) 1600MHz DDR3. Supported Operating Systems: Windows™ 7 Professional Service Pack 1 32-bit, Windows 10 Pro/IoT.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryCE Fragment Analysis (.fsa files)
For Use With (Application)Sequencing
LicenseClient
Operating SystemWindows 10 Pro/IoT, Windows 7 Pro
Product LineGeneMapper
Product TypeSoftware 6 Client Installation
Quantity1 License
SoftwareCE Fragment Analysis
Unit SizeEach
Contents & Storage
Software/CD
Frequently asked questions (FAQs)
How can I import Project Information into GeneMapper Manager?
Go to the Tools menu and select GeneMapper Manager. Click on the tab for the file(s) you want to import. Click on Import at the bottom of the window and select the files to be imported. Note: Multiple projects can be selected for importing at one time, but all other tabs are one at a time. Once imported, the new files will appear in the list. When finished, click Done in the lower right corner.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up my data files for DNA sequencing and fragment analysis applications, with the following extension, .ab1 or .fsa, on the capillary electrophoresis instruments?
The location of the sample files, .ab1 or .fsa, will either be in the Runs folder (D:/AppliedBio/310/Runs) on the ABI PRISM 310 Genetic Analyzer system or it will be determined by the Results Group in the Data Collection Software on the multi-capillary platforms. If you do not know where that location is, you can go to the Data Collection Software for your instrument and go to the Results Group section (top left corner in Unified Data Collection; for 3500 Data Collection, go to Library and in the Manage section, select Results Group), open the Results Group, and check the Sample File Folder location.
Save the files to the storage media of your choice (CD, DVD, network drive, flash drive).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do you have a manual uninstall procedure for Full Install configurations on a stand-alone computer for GeneMapper Software 5?
Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-software-5-uninstall%20-%20full-install.pdf) and follow the directions to uninstall GeneMapper Software 5.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I need to uninstall GeneMapper Software 5 Client because of failed installation. How can I do this for Client Install configurations on a stand-alone computer?
Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-5-uninstall-client-install.pdf) and follow the directions to uninstall GeneMapper Software 5.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
When installing the GeneMapper Software 5 Demo, I'm getting the message "Software requires tPKD Driver". What should I do?
Right-click on Computer and select Properties. In the System section, look at the System Type and see if you have Windows 7 Professional, 32-bit or 64-bit edition. Write this down, as you will need this information during the install process.
To install the necessary driver:
1. Open a browser and go to https://www.ilok.com/#!home.
2. On the right-hand side of the page, you should see the “Free Download License Support” links. If your computer has Windows 7 Professional, 32-bit edition, select the middle option (“Windows 32-bit XP, Vista, 7, 8”). If your computer has Windows 7 Professional, 64-bit edition, choose the third option (“Windows 64-bit XP, Vista, 7, 8”).
Note: If your browser gives you the choice to Run or Save, save the file. For the Internet Explorer browser, you can choose the location to save the file. For Mozilla Firefox and Google Chrome browsers, the default location to save the file is the Downloads folder.
3. Open the LicenseSupportInstallerWin zip file and go to the Old Installers Folder.
4. Run the InterLok Driver Setup Win (64 or 32).exe file.
You should now be able to install the GeneMapper Software 5 Demo.
If the problem persists, please contact Technical Support by sending an email to techsupport@thermofisher.com for further assistance.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.