Need help deciding what product to use with the Ion PGM™ System? Check out our helpful Applications Decision Tree for the Ion PGM™ System.
Product Image
Ion Torrent™

Ion AmpliSeq™ Cancer Hotspot Panel v2

The Ion AmpliSeq™Cancer Hotspot Panel v2 is a single pool of primers used to perform multiplex PCR for preparation ofRead more
Have Questions?
Catalog NumberQuantity
44753468 Reactions
Catalog number 4475346
Price (USD)
432.00
Each
Quantity:
8 Reactions
Price (USD)
432.00
Each

The Ion AmpliSeq™Cancer Hotspot Panel v2 is a single pool of primers used to perform multiplex PCR for preparation of amplicon libraries from genomic 'hot spot' regions that are frequently mutated in human cancer genes (download the list of targeted genes and mutations for the Ion AmpliSeq™Cancer Hotspot Panel v2). Building on the mutations included in our original Ion AmpliSeq™Cancer Panel, the latest Ion AmpliSeq™ready-to-use panel provides:

  • Maintained compatibility with FFPE samples while expanding mutational content for broader coverage of additional genes and 'hot spot' mutations
  • Extremely uniform coverage for more efficient sequencing and cost savings
  • Enhanced primer design with optimization of new primer sets and even lower strand bias for added confidence in accurate variant calling
  • Improved variant detection with Torrent Suite Software v3.0 and Variant Caller Plugin in low allele variant detection along with improved indel sensitivity

Additional Mutational Content for Advancing Cancer Research Studies

The Ion AmpliSeq™Cancer Hotspot Panel v2 is designed to amplify 207 amplicons covering approximately 2,800 COSMIC mutations from 50 oncogenes and tumor suppressor genes. This includes the 739 COSMIC mutations from 46 genes in the first Ion AmpliSeq™Cancer Panel along with added hotspot mutations from significant cancer genes. While maintaining our compatibility with FFPE samples and covering the same targets as the first Ion AmpliSeq™Cancer Panel, we have included additional mutations by adding several amplicons for hotspots in EZH2, GNA11, GNAQ, and IDH2, and by adding slightly longer amplicons for other genes to help advance your cancer research. The Ion AmpliSeq™Cancer Hotspot Panel v2 should be used with the Ion AmpliSeq™Library Kit 2.0.

More Efficient and More Cost-effective Sequencing with Extremely Uniform Coverage

The improved primer design in the Ion AmpliSeq™Cancer Hotspot Panel v2 improves upon the coverage uniformity of the Ion AmpliSeq™technology. With coverage that is more uniform, a minimum depth across all amplicons can be achieved using much less sequencing throughput. Consequently, additional samples can be multiplexed using any of the Ion chips. This results in cost savings with more efficient sequencing while maintaining high confidence in variant calling. Additionally, the low strand bias in the primer sets of the Ion AmpliSeq™Cancer Hotspot Panel v2 provides further confidence in accurate variant calling.

Simplicity, Speed, and Scalability of Ion AmpliSeq™Technology

Sustaining the simplicity of the Ion AmpliSeq™technology, the Ion AmpliSeq™Cancer Hotspot Panel v2 enables cancer genetic studies from FFPE tissues with as little as 10 ng of input DNA for targeted library construction. The Ion AmpliSeq™Cancer Hotspot Panel v2 also maintains the revolutionary workflow using standard PCR equipment and a simple PCR reaction for ultra-high multiplex PCR-based target selection, with no need for extensive capital investment. Additionally, the Ion AmpliSeq™Cancer Hotspot Panel v2 continues to enable you to achieve the fastest time to targeted libraries in approximately 3.5 hours. Scalability and flexibility is also preserved with the Ion AmpliSeq™Cancer Hotspot Panel v2, allowing the ability to sequence one sample or multiplex barcoded samples using any of the Ion chips.

Improved Variant Detection with Torrent Suite Software v3.0

The intuitive Torrent Suite Software and Variant Caller Plugin enable you to go from extracted DNA to variant calls in as little as 10 hours. With improvements to algorithms, you will see more sensitive indel detection along with more sensitive SNP detection with lower frequency alleles. Ion Reporter™Software can then be used for annotation of variants in routine DNA assays for the advancement of clinical research.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chip CompatibilityIon 520, Ion 530
For Use With (Application)Oncology Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
Multiplex Capability207 amplicons
No. of Reactions8
No. of SamplesUp to 8 Samples
Nucleic Acid TypeDNA
Product LineIon AmpliSeq
Product TypePanel
Quantity8 Reactions
Sample TypeFFPE Tissue, Blood, Cell Cultures, Clinical Samples, DNA (Genomic), DNA from FFPE, dsDNA, Fine Needle Aspirates (FNA), FFPE, Whole Blood
SpeciesHuman
Starting Material Amount10 ng DNA
TechniqueAmplicon Sequencing, Post-Light™ Ion Semiconductor Sequencing
ThroughputUp to 4 Samples per Ion 510 Chip, Up to 8 Samples per Ion 520 Chip, Up to 26 Samples per Ion 530 Chip
Workflow StepPanel
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

What is the difference between a DNA fragment library and a DNA amplicon library?

A DNA fragment library is constructed from whole genomic DNA and is commonly used for whole genome resequencing or de novo sequencing. Briefly, the whole genomic DNA is fragmented or sheared, ligated with Ion-specific adapter sequences, and then size-selected for the library fragments of the desired length.

Amplicon libraries are constructed from PCR-amplified DNA fragments and are used for targeted sequencing (e.g., investigating variants at known genomic locations). There are two types of amplicon libraries, short and long.

A short amplicon library contains DNA fragments (targets) with lengths that are compatible with the Ion template preparation kits without any further shearing or fragmentation during library preparation. Additionally, no size-selection step is required, as the amplicons are already within the desired size range.

A long amplicon library contains DNA fragments (targets) with lengths that are longer than those compatible with the Ion template kits and requires further shearing or fragmentation during library preparation. The library preparation protocol for long amplicons is similar to fragment libraries.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Is the Ion Library Equalizer Kit compatible with Ion Ampliseq libraries?

The Ion Library Equalizer Kit (Cat. No. 4482298) is recommended for use with Ion AmpliSeq libraries and provides an alternative to library quantification methods by using bead-based technology to normalize the final library concentration to ~100 pM.

The Ion Library Equalizer Kit is fast and cost-effective compared to traditional quantification methods; however, it may be not be the right choice for all users. Briefly, the library is amplified with the Ion Equalizer Primers, captured onto Equalizer Beads, and the normalized library is eluted from the beads using a specially formulated Equalizer Elution Buffer. The final library is normalized to ~100 pM, but there is no quality control information (e.g., measured concentration or size distribution) that can be obtained, which is possible if using the recommended library quantification kits: Ion Library Quantitation Kit (qPCR), Qubit dsDNA HS Assay Kit (Qubit 2.0 Fluorometer), or Agilent High Sensitivity DNA Kit (Agilent Bioanalyzer instrument).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What mutations are targeted in the Ion AmpliSeq Ready-to-Use and Community panels?

Detailed information regarding the variants targeted for each of the panels can be found at www.ampliseq.com. Under the Panels tab, find your panel of interest and press the “Download panel files” buttons. The ZIP archive will contain CSV files that include links to the COSMIC entries for each mutation on the cancer-related panels.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

My custom design has low predicted coverage, what can I do to improve it?

If you've reviewed your design and are not satisfied with the results, please click on the “Not happy with this design? Let us help” link to have an Ion Ampliseq team member contact you about additional options for your design.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How do I get started with the Ion AmpliSeq Designer?

Please visit the Ion AmpliSeq Designer website (www.ampliseq.com) and log in with your Thermofisher username and password. Select the “Help” tab to review training videos and documentation, including pipeline details, troubleshooting guides, and frequently asked questions.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.