DynaLight™ Substrate with RapidGlow™ Enhancer
Invitrogen™

DynaLight™ Substrate with RapidGlow™ Enhancer

Achieve faster ELISA results with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized toRead more
Have Questions?
Catalog NumberQuantity
4475406100 mL
Catalog number 4475406
Price (KRW)
1,061,000
Online offer
Ends: 30-Jun-2026
1,248,000
Save 187,000 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (KRW)
1,061,000
Online offer
Ends: 30-Jun-2026
1,248,000
Save 187,000 (15%)
Each
Achieve faster ELISA results with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized to achieve faster results in solution-based assays. The DynaLight™ Substrate with RapidGlow™ Enhancer formulation includes 1,2-dioxetane chemiluminescent substrate and a polymeric enhancer that enables ultra-sensitive immunoassay detection by alkaline phosphatase label.

Features of DynaLight™ Substrate with RapidGlow™ Enhancer:

• Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
• Sensitive attomole detection of purified alkaline phosphatase enzyme
• Optimized formulation for both microplate and bead-based assays
• Substrate stability of over 1 year at 4°C
• Compatibility with automated platforms with trigger solution/positions already in place

The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes at 37°C and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Assay Sensitivity1 pg/mL
Detection MethodChemiluminescence
For Use With (Equipment)Luminometer (Microplate)
FormLiquid
FormatBottle
High-throughput CompatibilityHigh-throughput Compatible
Product LineDynaLight, NovaBright, RapidGlow
Sample TypeCell Cultures, Whole Blood, Serum
Shipping ConditionWet Ice
Substrate PropertiesChemical Substrate
Substrate TypeAlkaline Phosphatase
Test Time2 min.
Product TypeELISA Substrate
Quantity100 mL
Unit SizeEach
Contents & Storage
1 amber bottle — Store at 2 to 8°C