5500 W FlowChip V2

Catalog number: 4475661

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 W FlowChip V2 is a microfluidic glass device with primers on the surface that enable amplification of library template onto the FlowChip for ligation sequencing on the 5500 W Series genetic analyzers. The 5500 W FlowChip V2 is specially designed with precisely cut lanes. Individual library samples can be deposited for amplification in each lane of the FlowChip. Sequencing reagents are delivered to each lane separately, enabling consumption of sequencing reagents for just the number of lanes used for sequencing.

The 5500 W FlowChip V2 is notched to set the orientation. Each 5500 W FlowChip V2 kit contains one 5500 W FlowChip V2 and a deposition cassette that aids in depositing sample libraries. The 5500 W FlowChip Prep Pack (available separately) contains reagents for depositing library template onto the 5500 W FlowChip V2.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Product Line: SOLiD™
Product Size: 1 kit
Type: Slides

Contents & storage

Store at 2°C to 8°C.

Documents

Manuals & protocols

Recommended products