5500 W FWD SR75 Kit

Catalog number: 4475669

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

5500 W FWD Sequencing Reagent (SR) kits contain the phosphatase and probes needed for ligation sequencing with 2-base encoded (2BE) probes on the 5500 W series genetic analyzers. The 5500 W FWD SR75 Kit consists of an 8-well striptube that contains sufficient phosphatase and probes to run up to 510 cycles and is best configured for sequencing read-lengths up to 75 bases with a 10-base barcode for 6 lanes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Recommended Applications: Resequencing, Short Read Sequencing
Product Line: Applied Biosystems™, SOLiD™
Product Size: 1 kit

Contents & storage

Store at -15 to -25 °C

Documents

Manuals & protocols

Recommended products