5500 W FWD2 SP Kit

Catalog number: 4475682

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 W FWD2 SP Kit contains sequencing primers and bridge probes for mate-paired sequencing runs, as well as multiplex fragment and multiplex paired-end sequencing runs. These are used to prime (1) the R3 tag of mate-paired libraries or (2) the barcode tag of multiplexed libraries for ligation sequencing in the forward direction.

The 5500 W FWD2 SP Kit includes:
1 strip tube containing specific sequencing primers and bridge probes
1 5500 W Sequencing QC strip tube
4 strip tubes of 5500 W mixing tubes

The kit contains sufficient primers to run 6 lanes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Applied Biosystems™, SOLiD™
Product Size: 1 kit
Shipping Condition: Dry Ice

Contents & storage

Store at -15 to -25 °C

Documents

Manuals & protocols