5500 W Buffer A

Catalog number: 4475690

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

5500 W Buffer A is needed for ligation sequencing on the 5500 W Series genetic analyzers. 5500 W Buffer A consists of 5500 W Buffer A Part 1 and 5500 W Buffer A Part 2, which need to be mixed prior to running on the instrument. 5500 W Buffer A contains sufficient reagents to run up to 432 cycles, or 72 cycles per lane for 6 lanes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Product Line: SOLiD™
Product Size: 1 kit
Shipping Condition: Wet Ice

Contents & storage

Store at 2 to 8 °C

Documents

Manuals & protocols