5500 W FlowChip Preparation Kit

Catalog number: 4475695

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 W FlowChip Preparation Kit contains reagents required to deposit sample library template into the 5500 W FlowChip and prepare for template amplification on the 5500 W Series genetic analyzers. This kit is an individually orderable component of the 5500 W FlowChip Prep Pack.

The 5500 W FlowChip Preparation Kit includes:
5500 W Template Wash Buffer
5500 W Hyb Solution

The kit contains sufficient reagents for 6 lanes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Recommended Applications: Resequencing, Short Read Sequencing
Product Line: SOLiD™
Product Size: 1 kit
Technique: Ligation-Based Sequencing
Shipping Condition: Room Temperature

Contents & storage

Store at ambient temperature

Documents

Manuals & protocols