Gallic acid monohydrate, ACS reagent
Gallic acid monohydrate, ACS reagent
Gallic acid monohydrate, ACS reagent
Thermo Scientific Chemicals

Gallic acid monohydrate, ACS reagent

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 447581000
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.80
Save 14.15
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.80
Save 14.15
Each
Chemical Identifiers
CAS5995-86-8
IUPAC Name3,4,5-trihydroxybenzoate
Molecular FormulaC7H5O5
InChI KeyLNTHITQWFMADLM-UHFFFAOYSA-M
SMILESOC1=CC(=CC(O)=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Insoluble matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only