Xylenol orange, sodium salt, ACS reagent
Xylenol orange, sodium salt, ACS reagent
Xylenol orange, sodium salt, ACS reagent
Thermo Scientific Chemicals

Xylenol orange, sodium salt, ACS reagent

CAS: 3618-43-7 | C31H28N2Na4O13S | 760.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 447600250
Price (USD)
326.65
Special offer
Online exclusive
Ends: 28-Dec-2026
384.00
Save 57.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
326.65
Special offer
Online exclusive
Ends: 28-Dec-2026
384.00
Save 57.35
Each
Chemical Identifiers
CAS3618-43-7
IUPAC Nametetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular FormulaC31H28N2Na4O13S
InChI KeyFXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Suitability(for Zn titration) Passes test
Appearance (Color)Red-brown to dark brown
Appearance (Form)Crystalline powder
Clarity of solutionPasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only