Attune™ Flow Cytometer Syringe
Actual product may vary
Applied Biosystems™

Attune™ Flow Cytometer Syringe

The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flowRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
447609010 mL
44520791 mL
Catalog number 4476090
Price (USD)
1,350.65
Online Exclusive
1,422.00
Save 71.35 (5%)
Each
Quantity:
10 mL
Price (USD)
1,350.65
Online Exclusive
1,422.00
Save 71.35 (5%)
Each

The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell. Replacement of this syringe is recommended as part of instrument maintenance.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity10 mL
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Flow Cytometer
Product TypeSyringe
Unit SizeEach
Contents & Storage
Store at room temperature