SOLiD™ Buffer F, Double, v2

Catalog number: 4476391

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SOLiD™ buffers are needed for ligation sequencing on the 5500/5500xl Genetic Analyzer. These buffers are individually orderable components of the SOLiD™ Run Cycle Buffer Kit. This double size buffer has sufficient quantity for two FlowChips.

SOLiD™ Buffer F, Double, v2 contains sufficient reagents to run 950 to 1632 cycles or up to 136 cycles per lane. SOLiD™ Buffer F, Double, v2 comes at a 10X concentration and must be diluted to 1X before use.

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.

Specifications

Buffer Type: Sequencing Buffer
Concentrated: 10 X
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Form: Liquid
Product Line: SOLiD™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols