5500 W Run Cycle Buffer Kit

Catalog number: 4476801

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

5500 W buffers are used for ligation sequencing on the 5500 W Series genetic analyzers. The 5500 W Run Cycle Buffer Kit contains a set of six buffers required during all types of sequencing runs for ligation sequencing. The kit consists of 5500 W Buffers A, B, C, E, F, and G. The 5500 W Run Cycle Buffer Kit does not contain 5500 W Buffer D or 5500 W FWD Buffer.

5500 W Buffers A, B, and E are provided in two parts, and components must be mixed before use. 5500 W Buffer F is provided as a 10X solution and requires dilution to 1X before pouring into its reagent tank. 5500 W Buffer G is poured into its reagent tank before use.

The kit contains sufficient reagents to run up to 432 cycles, or 72 cycles per lane.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Sequencing Buffer
For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Product Line: Applied Biosystems™, SOLiD™
Product Size: 1 kit

Contents & storage

Store at -15 to -25 °C, 2 to 8 °C, and ambient

Documents

Manuals & protocols