Thermo Scientific Chemicals

Citric acid, diammonium salt, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447681000
100 g, Each
Thermo Scientific Chemicals

Citric acid, diammonium salt, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447681000
100 g, Each
Quantity

Chemical Identifiers

CAS
3012-65-5
IUPAC Name
diammonium 3-carboxy-3-hydroxypentanedioate
Molecular Formula
C6H14N2O7
InChI Key
YXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
Appearance (Form)
Crystalline powder
Titration with NaOH
98.0 to 103.0 %
Heavy metals (ICP-OES)
=<5 ppm
Iron (Fe)
=<0.001 %
Oxalate (C2O4)
Passes test (Limit about 0.05%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text