Thermo Scientific Chemicals

Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 447712500
250 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 447712500
250 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 447712500
Price (USD)
Price: 86.50
Online price: 73.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6108-17-4
IUPAC Name
lithium(1+) acetate dihydrate
Molecular Formula
C2H7LiO4
InChI Key
IAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES
[Li+].O.O.CC([O-])=O
Appearance (Form)
Crystalline powder
Titration with HClO4
97.5 to 102.5 %
Insoluble matter
=<0.01 % (in water)
pH
7.5 to 9.5 (5 % in water)
Chloride (Cl)
=<0.003 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text